Learn More
HDAC3 Mouse anti-Human, Clone: OTI4E10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806191

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. HDAC3 belongs to the histone deacetylase/acuc/apha family. It has histone deacetylase activity and represses transcription when tethered to a promoter. It may participate in the regulation of transcription through its binding with the zinc-finger transcription factor YY1. This protein can also down-regulate p53 function and thus modulate cell growth and apoptosis. This gene is regarded as a potential tumor suppressor gene.Specifications
HDAC3 | |
Monoclonal | |
Unconjugated | |
HDAC3 | |
HD3, RPD3, RPD3-2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
8841 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4E10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O15379 | |
HDAC3 | |
Human recombit protein fragment corresponding to amino acids 1-235 of human HDAC3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.