Learn More
Invitrogen™ HDAC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA1862
Catalog No. PA1862
Only null left
Edge
Add to Cart
Description
Description
PA1-862 detects histone deacetylase 3 (HDAC3) from canine, hamster, human and rat tissues and cells. PA1-862 has been successfully used in Western blot and ChIP procedures. By Western blot, this antibody detects an ~48 kDa protein representing HDAC3 from HeLa cell lysate. The PA1-862 immunogen is a synthetic peptide corresponding to residues C D(415) G D H D N D K E S D V E I(428) of human HDAC3. This immunizing peptide (Cat. # PEP-071) is available for use in neutralization and control experiments.
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. HDAC3 belongs to the histone deacetylase/acuc/apha family. It has histone deacetylase activity and represses transcription when tethered to a promoter. It may participate in the regulation of transcription through its binding with the zinc-finger transcription factor YY1. This protein can also down-regulate p53 function and thus modulate cell growth and apoptosis. This gene is regarded as a potential tumor suppressor gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HDAC3
Polyclonal
Unconjugated
HDAC3
AW537363; HD3; Hdac3; HDAC3 splicing HDAC3alpha; HDAC3 splicing HDAC3beta; HDAC3 splicing HDAC3delta; HDAC3 splicing HDAC3gamma; histone deacetylase 3; histone deacetylase 3-like protein; RPD3; RPD3-2; SMAP45
Rabbit
Antigen affinity chromatography
RUO
100752639, 478040, 84578, 8841
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 1 mg/mL BSA and 0.05% sodium azide
O15379, Q6P6W3
HDAC3
Synthetic peptide corresponding to residues C D(415) G D H D N D K E S D V E I(428) of human HDAC3.
100 μg
Primary
Human, Rat, Canine, Hamster
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.