Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC3 Rabbit anti-Human, Mouse, Rat, Clone: 1M1O7, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | HDAC3 |
---|---|
Clone | 1M1O7 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
HDAC3 Monoclonal antibody specifically detects HDAC3 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
HDAC3 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
8841 | |
IgG | |
Affinity purified |
1M1O7 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45 | |
A synthetic peptide corresponding to a sequence within amino acids 329-428 of human HDAC3 (O15379). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title