Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726155 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6155 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6155 Supplier Proteintech Group Inc Supplier No. 102551AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is a rabbit polyclonal antibody raised against an internal region of human HDAC3.

Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4), and some other non-histone substrates. Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Participates in the BCL6 transcriptional repressor activity by deacetylating the H3 ′Lys-27′ (H3K27) on enhancer elements, antagonizing EP300 acetyltransferase activity and repressing proximal gene expression. Probably participates in the regulation of transcription through its binding to the zinc-finger transcription factor YY1; increases YY1 repression activity. Required to repress transcription of the POU1F1 transcription factor. Acts as a molecular chaperone for shuttling phosphorylated NR2C1 to PML bodies for sumoylation. Contributes, together with XBP1 isoform 1, to the activation of NFE2L2-mediated HMOX1 transcription factor gene expression in a PI(3)K/mTORC2/Akt-dependent signaling pathway leading to endothelial cell (EC) survival under disturbed flow/oxidative stress (By similarity).

Specifications

Antigen HDAC3
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene HDAC3
Gene Accession No. O15379, O88895, Q6P6W3
Gene Alias HD3, HDAC3, histone deacetylase 3, RPD3, RPD3 2, SMAP45
Gene Symbols Hdac3
Host Species Rabbit
Immunogen HDAC3 Fusion Protein Ag0396
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15183, 84578, 8841
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.