Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDAC3 Recombinant Rabbit Monoclonal Antibody (1M1O7)
GREENER_CHOICE

Catalog No. PIMA542703
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542703 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542703 Supplier Invitrogen™ Supplier No. MA542703
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: PC-3, HeLa, Mouse brain, NIH/3T3, Rat testis. The target is usually found in the following locations: Cytoplasm, Nucleus, cytosol. Immunogen sequence: FEYFAPDFTL HPDVSTRIEN QNSRQYLDQI RQTIFENLKM LNHAPSVQIH DVPADLLTYD RTDEADAEER GPEENYSRPE APNEFYDGDH DNDKESDVEI.

Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. HDAC3 belongs to the histone deacetylase/acuc/apha family. It has histone deacetylase activity and represses transcription when tethered to a promoter. It may participate in the regulation of transcription through its binding with the zinc-finger transcription factor YY1. This protein can also down-regulate p53 function and thus modulate cell growth and apoptosis. This gene is regarded as a potential tumor suppressor gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC3
Applications ELISA, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 1M1O7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HDAC3
Gene Accession No. O15379, O88895, Q6P6W3
Gene Alias AW537363; HD3; Hdac3; HDAC3 splicing HDAC3alpha; HDAC3 splicing HDAC3beta; HDAC3 splicing HDAC3delta; HDAC3 splicing HDAC3gamma; histone deacetylase 3; histone deacetylase 3-like protein; RPD3; RPD3-2; SMAP45
Gene Symbols HDAC3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 329-428 of human HDAC3 (NP_0038742).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15183, 84578, 8841
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.