Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC3 Recombinant Rabbit Monoclonal Antibody (8H2L16)
HDAC3 Recombinant Rabbit Monoclonal Antibody (8H2L16)
Supplier: Thermo Scientific 702440
Description
HDAC3 Recombinant Monoclonal Antibody for Western Blot

Specifications
HDAC3 | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
O15379, O88895, Q6P6W3 | |
HDAC3 | |
Protein corresponding to human HDAC3. | |
100 μg | |
Primary | |
Human, Mouse, Rat, Cynomolgus Monkey | |
Antibody | |
IgG |
Western Blot | |
8H2L16 | |
Unconjugated | |
HDAC3 | |
AW537363; HD3; Hdac3; HDAC3 splicing HDAC3alpha; HDAC3 splicing HDAC3beta; HDAC3 splicing HDAC3delta; HDAC3 splicing HDAC3gamma; histone deacetylase 3; histone deacetylase 3-like protein; RPD3; RPD3-2; SMAP45 | |
Rabbit | |
Protein A | |
RUO | |
102119873, 15183, 84578, 8841 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction