Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDAC4 Monoclonal Antibody (7B2)
GREENER_CHOICE

Catalog No. PIMA515580
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515580 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515580 Supplier Invitrogen™ Supplier No. MA515580
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA5-15580 targets HDAC4 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15580 immunogen is purified recombinant fragment of human HDAC4 expressed in E. Coli. MA5-15580 detects HDAC4 which has a predicted molecular weight of approximately 119 kDa.

Histone deacetylase 4 (HDAC4) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3, and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. HDAC4 is also involved in muscle maturation via its interaction with the myocyte enhancer factors such as MEF2A, MEF2C and MEF2D. It is also involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer. Mutations to the HDAC4 gene have been found in Brachydactyly-mental retardation syndrome (BDMR).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC4
Applications ChIP Assay, ELISA, Western Blot
Classification Monoclonal
Clone 7B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene HDAC4
Gene Accession No. P56524, Q6NZM9
Gene Alias 4932408F19Rik; AHO3; AI047285; BDMR; DKFZp779K1053; HA6116; HD4; HD7; HD7b; HD9; HDAC; HDAC4; HDAC-4; HDAC7; HDAC7B; HDAC9; HDAC9B; HDAC9FL; HDACA; HDAC-A; HDRP; Histone deacetylase 4; histone deacetylase A; KIAA0288; KIAA0744; MITR
Gene Symbols HDAC4
Host Species Mouse
Immunogen Purified recombinant fragment of human HDAC4 expressed in E. Coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208727, 9759
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.