Learn More
HDAC4 Mouse anti-Human, Clone: OTI4A4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805010

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Histone deacetylase 4 (HDAC4) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3, and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. HDAC4 is also involved in muscle maturation via its interaction with the myocyte enhancer factors such as MEF2A, MEF2C and MEF2D. It is also involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer. Mutations to the HDAC4 gene have been found in Brachydactyly-mental retardation syndrome (BDMR).Specifications
HDAC4 | |
Monoclonal | |
Unconjugated | |
HDAC4 | |
AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA | |
Mouse | |
Affinity Chromatography | |
RUO | |
9759 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4A4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P56524 | |
HDAC4 | |
Full length human recombit protein of human HDAC4 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.