Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HDAC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581191
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human HDAC4.
Histone deacetylase 4 (HDAC4) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3, and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. HDAC4 is also involved in muscle maturation via its interaction with the myocyte enhancer factors such as MEF2A, MEF2C and MEF2D. It is also involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer. Mutations to the HDAC4 gene have been found in Brachydactyly-mental retardation syndrome (BDMR).
Specifications
HDAC4 | |
Polyclonal | |
Unconjugated | |
HDAC4 | |
4932408F19Rik; AHO3; AI047285; BDMR; DKFZp779K1053; HA6116; HD4; HD7; HD7b; HD9; HDAC; HDAC4; HDAC-4; HDAC7; HDAC7B; HDAC9; HDAC9B; HDAC9FL; HDACA; HDAC-A; HDRP; Histone deacetylase 4; histone deacetylase A; KIAA0288; KIAA0744; MITR | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
9759 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with no preservative | |
P56524 | |
HDAC4 | |
A synthetic peptide corresponding to the N-terminus of the Human HDAC4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction