Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 161651AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-0764


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

This antibody is a rabbit polyclonal antibody raised against a peptide mapping within human HDAC4.

Histone deacetylase 4 (HDAC4) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3, and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. HDAC4 is also involved in muscle maturation via its interaction with the myocyte enhancer factors such as MEF2A, MEF2C and MEF2D. It is also involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer. Mutations to the HDAC4 gene have been found in Brachydactyly-mental retardation syndrome (BDMR).
Specifications

Specifications

HDAC4
Polyclonal
Unconjugated
HDAC4
HA6116, HD4, HDAC A, HDAC4, HDACA, histone deacetylase 4, KIAA0288
Rabbit
Antigen Affinity Chromatography
RUO
208727, 363287, 9759
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P56524, Q6NZM9, Q99P99
Hdac4
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.