Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC6 Antibody (236), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305295
This item is not returnable.
View return policy
Description
HDAC6 Monoclonal antibody specifically detects HDAC6 in Human, Fish (Negative), Mouse (Negative) samples. It is validated for Western Blot, ELISA, Immunofluorescence, ImmunoprecipitationSpecifications
| HDAC6 | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Fish (Negative), Mouse (Negative) |
| Western Blot, ELISA, Immunofluorescence, Immunoprecipitation | |
| 236 | |
| Western Blot 1-5 ug/mL, ELISA, Immunocytochemistry/Immunofluorescence 5-10 ug/mL, Immunoprecipitation | |
| EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
| Human HDAC6 | |
| 0.1 mg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Neuroscience | |
| 10013 | |
| Store at 4°C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction