Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC6 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178981C
Description
HDAC6 Polyclonal specifically detects HDAC6 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot (Negative), Immunohistochemistry (Negative).Specifications
HDAC6 | |
Polyclonal | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence 1:100, Western Blot (Negative), Immunohistochemistry (Negative) | |
Q9UBN7 | |
HDAC6 | |
A synthetic peptide made to a C-terminal portion of the human HDAC6 protein (between residues 1175-1215). [UniProt Q9UBN7] | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases, Neuroscience | |
10013 | |
Human, Mouse | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human and mouse. Mouse data from customer feedback. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction