Learn More
HDAC6 (internal)Rabbit anti-Human, Polyclonal, Rockland™

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 600401J39

Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. A BLAST analysis was used to suggest cross-reactivity with monkey and human based on 100% sequence homology. Cross-reactivity with HDAC6 from other sources has not been determined.
Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.Specifications
HDAC6 (internal) | |
Polyclonal | |
Unconjugated | |
HDAC6 | |
CPBHM; FLJ16239; HD6; HDA6; Hdac5; HDAC6; histone deacetylase 5; Histone deacetylase 6; Histone deacetylase mHDA2; JM21; KIAA0901; mHDA2; PPP1R90; protein phosphatase 1, regulatory subunit 90; scurfy candidate 6; Sfc6 | |
Rabbit | |
RUO | |
10013 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1.09 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl, 30% glycerol and 0.05% sodium azide; pH 7.2 | |
Q9UBN7 | |
HDAC6 | |
100 μg | |
Primary | |
Human | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.