Learn More
Invitrogen™ HDAC6 Monoclonal Antibody (236)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA562374

Description
HDAC6 Monoclonal Antibody for Western Blot, ICC/IF, Flow
Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.
Specifications
HDAC6 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Q9UBN7 | |
HDAC6 | |
Human HDAC6. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Flow Cytometry, Western Blot, Immunocytochemistry | |
236 | |
Unconjugated | |
HDAC6 | |
CPBHM; FLJ16239; Hd6; HDA6; Hdac5; Hdac6; histone deacetylase 5; histone deacetylase 6; Histone deacetylase mHDA2; JM21; KIAA0901; mHDA2; PPP1R90; protein phosphatase 1, regulatory subunit 90; scurfy candidate 6; Sfc6 | |
Mouse | |
Protein A | |
RUO | |
10013 | |
4°C, do not freeze | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.