Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDAC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595908 Supplier Invitrogen™ Supplier No. PA595908
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTSTGQDSTT TRQRRSRQNP QSPPQDSSVT SKRNIKKGAV PRSIPNLAEV KKKGKMKKLG QAMEEDLIVG LQGMDLNLEA EALAGTGLVL DEQLNEFHCL WDDSFPEGPE RLHAIKEQLI QEGLLDRCVS FQARFAEKEE LMLVHSLEYI DLMETTQYMN EGELRVLADT YDSVYLHPNS YSCACLASGS VLRLVDAVLG; Positive Samples: K-562, HeLa; Cellular Location: Cell projection, Cytoplasm, Nucleus, Perikaryon, axon, dendrite.

Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HDAC6
Gene Accession No. Q9UBN7, Q9Z2V5
Gene Alias CPBHM; FLJ16239; Hd6; HDA6; Hdac5; Hdac6; histone deacetylase 5; histone deacetylase 6; Histone deacetylase mHDA2; JM21; KIAA0901; mHDA2; PPP1R90; protein phosphatase 1, regulatory subunit 90; scurfy candidate 6; Sfc6
Gene Symbols HDAC6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human HDAC6 (NP_0060352).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10013, 15185, 84581
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.