Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC6 Rabbit anti-Human, Mouse, HRP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10056343H
Description
HDAC6 Polyclonal specifically detects HDAC6 in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
HDAC6 | |
Polyclonal | |
Western Blot, Simple Western | |
EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
Rabbit | |
Protein G purified | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases, Neuroscience | |
10013.0 | |
Human, Mouse | |
IgG |
Western Blot | |
HRP | |
PBS with No Preservative | |
HDAC6 | |
Synthetic peptide made to an N-terminal portion of human HDAC6 (between amino acids 1-50) [UniProt Q9UBN7] | |
0.1 mL | |
Primary | |
In NIH 3T3, a 134 kDa band is observed. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction