Learn More
HDAC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 161671AP150UL

Description
This antibody is specific to HDAC6. It does not cross react with other HDACs.
Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.Specifications
HDAC6 | |
Polyclonal | |
Unconjugated | |
HDAC6 | |
HD6, HDAC6, histone deacetylase 6, JM21, KIAA0901 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10013, 15185, 84581 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UBN7, Q9Z2V5 | |
Hdac6 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.