Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDAC6 Recombinant Rabbit Monoclonal Antibody (2S1H4)
GREENER_CHOICE

Catalog No. FEMA551331
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551331 Supplier Invitrogen™ Supplier No. MA551331
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: NGAAVVRPPG HHAEQDAACG FCFFNSVAVA ARHAQTISGH ALRILIVDWD VHHGNGTQHM FEDDPSVLYV SLHRYDHGTF FPMGDEGASS QIGRAAGTGF T.

Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC6
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 2S1H4
Concentration 0.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene HDAC6
Gene Accession No. Q9UBN7
Gene Alias CPBHM; FLJ16239; Hd6; HDA6; Hdac5; Hdac6; histone deacetylase 5; histone deacetylase 6; Histone deacetylase mHDA2; JM21; KIAA0901; mHDA2; PPP1R90; protein phosphatase 1, regulatory subunit 90; scurfy candidate 6; Sfc6
Gene Symbols HDAC6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 600-700 of human HDAC6 (NP_006035.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10013
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.