Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC7 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298759PCP
Description
HDAC7 Polyclonal antibody specifically detects HDAC7 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
HDAC7 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HDAC7. | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Phospho Specific | |
51564 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction