Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC7, p Ser155 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$243.50 - $341.50

Specifications

Antigen HDAC7 (p Ser155)
Concentration 0.5 mg/mL
Dilution Western Blot 0.1-1 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224454SS
SDP
View Documents
Novus Biologicals
NBP224454SS
0.025 mg
Each for $243.50
Only null left
Add to Cart
 
NBP224454
SDP
View Documents
Novus Biologicals
NBP224454
0.05 mg
Each for $341.50
Only null left
Add to Cart
 
Description

Description

HDAC7 Polyclonal specifically detects HDAC7 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot.
Specifications

Specifications

HDAC7 (p Ser155)
Western Blot 0.1-1 ug/ml
Polyclonal
Rabbit
Breast Cancer, Cancer, Chromatin Research, Phospho Specific
0.1 ml PBS and 0.05% BSA with 0.05% Sodium Azide
51564
A synthetic peptide containing phospho-serine 155 of human HDAC-7 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
RUO
Human, Mouse, Rat, Bovine, Canine, Primate
DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917
HDAC7
IgG
Affinity Purified
The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, and dog, 94% homologous in chicken, opossum and pufferfish and 83% homologous in zebrafish.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.