Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC7, p Ser448 Antibody, Novus Biologicals™
SDP

Catalog No. p-5071231 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP203979SS 0.025 mg
NBP203979 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP203979SS Supplier Novus Biologicals Supplier No. NBP203979SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HDAC7 Polyclonal specifically detects HDAC7 in Human, Rat, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot.

Specifications

Antigen HDAC7 (p Ser448)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.1-0.5 ug/ml
Formulation 0.1 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. NP_055522
Gene Alias DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917
Gene Symbols HDAC7
Host Species Rabbit
Immunogen Within the range of amino acids 443-460 of human HDAC-7 protein were used as the immunogen ('b' represents a phosphorylated serine at position 448).
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 51564
Test Specificity Reacts with Chimpanzee, Cow, Dog, Horse, Human, New World Monkey and Rat. Predicted to react with Mouse. The amino acid sequence used as immunogen is 100% homologous in Human, Monkey, Cow, Dog, Chimpanzee and Horse and 90% homologous in Mouse.
Target Species Human, Rat, Bovine, Canine, Equine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.