Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC9 Mouse anti-Human, Clone:2B7C4, Abnova™

Catalog No. 89333097 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-097 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-097 Supplier Abnova Corporation Supplier No. MAB16732
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human HDAC9.

Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. The protein encoded by this gene has sequence homology to members of the histone deacetylase family. This gene is orthologous to the Xenopus and mouse MITR genes. The MITR protein lacks the histone deacetylase catalytic domain. It represses MEF2 activity through recruitment of multicomponent corepressor complexes that include CtBP and HDACs. This encoded protein may play a role in hematopoiesis. Multiple alternatively spliced transcripts have been described for this gene but the full-length nature of some of them has not been determined. [provided by RefSeq]

Specifications

Antigen HDAC9
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2B7C4
Conjugate Unconjugated
Description histone deacetylase 9
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene HDAC9
Gene Alias DKFZp779K1053, HD7, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, KIAA0744, MITR
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 343-569 of human HDAC9 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 9734
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.