Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC9 Mouse anti-Human, Clone: OTI7G2, lyophilized, TrueMAB™
SDP

Catalog No. 501667384
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-7384 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-7384 Supplier OriGene Supplier No. CF805328
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

HDAC-9 is a histone deacetylase with an important role in the regulation of gene expression and hematopoiesis. A nuclear protein, HDAC-9 belongs to the histone deacetylase family with several isoforms and is inhibited by Trichostatin A (TSA) and suberoylanilide hydroxamic acid. It helps in deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4) and gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. MEF2-dependent transcription is repressed by HDAC-9 by recruiting HDAC-1 and/or HDAC-3. It also inhibits skeletal myogenesis and is involved in heart development. Ubiquitous but variable expression is seen in brain and other tissues like heart, pancreas and also in human bladder carcinoma cells. Lack of HDAC-9 protein is found in a family with Peters anomaly, a central corneal leukoma.

Specifications

Antigen HDAC9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7G2
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene HDAC9
Gene Accession No. Q9UKV0
Gene Alias HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Gene Symbols HDAC9
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 181-460 of human HDAC9 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9734
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.