Learn More
Invitrogen™ HDBP2 Polyclonal Antibody

Description
Sequence homology: Cow: 85%; Dog: 92%; Guinea Pig: 79%; Horse: 92%; Human: 100%; Mouse: 92%; Rat: 92%.
Specifications
Specifications
| Antigen | HDBP2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | ZNF395 |
| Gene Accession No. | Q9H8N7 |
| Gene Alias | BC053701; Gm910; HD gene regulatory region-binding protein 2; HDBP2; HDBP-2; HD-regulating factor 2; HDRF-2; Huntington disease gene regulatory region-binding protein 2; Huntington's disease gene regulatory region-binding protein 2; papillomavirus regulatory factor; papillomavirus regulatory factor 1; papillomavirus regulatory factor PRF-1; papillomavirus-binding factor; PBF; PRF1; PRF-1; Si-1-8-14; Zfp395; Zinc finger protein 395; ZNF395 |
| Gene Symbols | ZNF395 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.