Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550675
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550675 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550675 Supplier Invitrogen™ Supplier No. PA550675
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ZNF395 protein.

Papillomavirus Regulatory Factor 1/HDBP2 is a novel transcription factor shuttling between nucleus and cytoplasm and binds to the specific GCCGGCG, which is an essential cis-element for Huntington's disease gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDBP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ZNF395
Gene Accession No. Q9H8N7
Gene Alias BC053701; Gm910; HD gene regulatory region-binding protein 2; HDBP2; HDBP-2; HD-regulating factor 2; HDRF-2; Huntington disease gene regulatory region-binding protein 2; Huntington's disease gene regulatory region-binding protein 2; papillomavirus regulatory factor; papillomavirus regulatory factor 1; papillomavirus regulatory factor PRF-1; papillomavirus-binding factor; PBF; PRF1; PRF-1; Si-1-8-14; Zfp395; Zinc finger protein 395; ZNF395
Gene Symbols ZNF395
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human zinc finger protein 395.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55893
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.