Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HDC Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | HDC |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119194
|
Invitrogen™
PA5119194 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121918
|
Invitrogen™
PA5121918 |
20 μL |
Each for $209.50
|
|
|||||
Description
HDC Polyclonal Antibody for Western Blot, IHC (P)
The biogenic amine histamine is an important modulator of numerous physiologic processes, including neurotransmission, gastric acid secretion, and smooth muscle tone. The biosynthesis of histamine from histidine is catalyzed by the enzyme L-histidine decarboxylase (HDC; EC 4.1.1.22). This homodimeric enzyme is a pyridoxal phosphate (PLP)-dependent decarboxylase and is highly specific for its histidine substrate.Specifications
HDC | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
Hdc | |
AW108189; HDC; Hdc-a; Hdc-c; Hdc-e; Hdc-s; histidine decarboxylase; L-histidine decarboxylase; MGC163399 | |
Hdc | |
Primary | |
Antigen affinity chromatography |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P19113 | |
3067 | |
Recombinant protein Histidine Decarboxylase. The antigen corresponds to amino acid range 160-369 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title