Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDC Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980927
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0927 Supplier Bioss Supplier No. BS1054R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The biogenic amine histamine is an important modulator of numerous physiologic processes, including neurotransmission, gastric acid secretion, and smooth muscle tone. The biosynthesis of histamine from histidine is catalyzed by the enzyme L-histidine decarboxylase (HDC; EC 4.1.1.22). This homodimeric enzyme is a pyridoxal phosphate (PLP)-dependent decarboxylase and is highly specific for its histidine substrate.

Specifications

Antigen HDC
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Hdc
Gene Accession No. P19113
Gene Alias AW108189; Hdc; Hdc-a; Hdc-c; Hdc-e; Hdc-s; Histidine decarboxylase; L-histidine decarboxylase; MGC163399
Gene Symbols Hdc
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HDC.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3067
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.