Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDDC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557363
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557363 Supplier Invitrogen™ Supplier No. PA557363
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASVSSATFS GHGARSLLQF LLLVGQLKRV PRTGWVYRNV QRPESVSDHM YRMAVMAMVI KDDRLNKDRC VRLALVHDMA ECIVGDIAPA DNIPKEEKHR REEEAMKQIT QLLP Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

Catalyzes the dephosphorylation of the nucleoside 5'-monophosphates deoxyadenosine monophosphate (dAMP), deoxycytidine monophosphate (dCMP), deoxyguanosine monophosphate (dGMP) and deoxythymidine monophosphate (dTMP). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDDC2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Hddc2
Gene Accession No. Q7Z4H3
Gene Alias 2310057G13Rik; C6orf74; CGI-130; dJ167O5.2; HCV NS5A-transactivated protein 2; HD domain containing 2; HD domain-containing protein 2; HDDC2; Hepatitis C virus NS5A-transactivated protein 2; NS5ATP2; RGD1307562; testicular tissue protein Li 83
Gene Symbols Hddc2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HDDC2. Recombinant protein control fragment (Product #RP-94654).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51020
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.