Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDDC3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156806
Description
HDDC3 Polyclonal specifically detects HDDC3 in Human samples. It is validated for Western Blot.Specifications
| HDDC3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| HDDC3 | |
| Synthetic peptides corresponding to HDDC3(HD domain containing 3) The peptide sequence was selected from the middle region of HDDC3. Peptide sequence TDDKTLPKLERKRLQVEQAPHSSPGAKLVKLADKLYNLRDLNRCTPEVKI. | |
| 100 μL | |
| Proteases & Other Enzymes | |
| 374659 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| (ppGpp)ase, EC 3.1.7.2, guanosine-3'-5'-bis(diphosphate) 3'-pyrophosphohydrolase MESH1, HD domain containing 3, HD domain-containing protein 3, MESH1, Metazoan SpoT homolog 1, metazoan SpoT homolog-1, MGC45386, Penta-phosphate guanosine-3'-pyrophosphohydrolase | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Human: 100%; Pig: 100%; Zebrafish: 100%; Rat: 92%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction