Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDGF Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171924AF700
Description
HDGF Polyclonal antibody specifically detects HDGF in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
HDGF | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
FLJ96580, hepatoma-derived growth factor, hepatoma-derived growth factor (high-mobility group protein 1-like), High mobility group protein 1-like 2, HMG-1L2, HMG1L2DKFZp686J1764 | |
The immunogen for this product maps to a region between residue 100 and 150 of human Hepatoma-Derived Growth Factor using the numbering given in entry NP_004485.1 (GeneID 3068). | |
0.1 mL | |
Cancer, Cell Biology | |
3068 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction