Learn More
HDGF Mouse anti-Human, Clone: OTI8C7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808926

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the hepatoma-derived growth factor family. The encoded protein has mitogenic and DNA-binding activity and may play a role in cellular proliferation and differentiation. This gene was thought initially to be located on chromosome X, however, that location has been determined to correspond to a related pseudogene. Alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
HDGF | |
Monoclonal | |
Unconjugated | |
HDGF | |
HMG1L2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3068 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8C7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P51858 | |
HDGF | |
Full length human recombit protein of human HDGF produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.