Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDGF Mouse anti-Human, Mouse, Rat, Clone: 5A7G3, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 600641IG150UL

Description
This gene encodes a member of the hepatoma-derived growth factor family. The encoded protein has mitogenic and DNA-binding activity and may play a role in cellular proliferation and differentiation. This gene was thought initially to be located on chromosome X, however, that location has been determined to correspond to a related pseudogene.Specifications
HDGF | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P51858, P51859, Q8VHK7 | |
Hdgf | |
HDGF Fusion Protein Ag1893 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
5A7G3 | |
Unconjugated | |
HDGF | |
HDGF, Hepatoma derived growth factor, HMG 1L2, HMG1L2 | |
Mouse | |
Protein G | |
RUO | |
114499, 15191, 3068 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction