Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDGF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561810 Supplier Invitrogen™ Supplier No. PA561810
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEGEEKEAAT LEVERPLPME VEKNSTPSEP GS Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 75%.

HDGF is a member of the hepatoma-derived growth factor family. This protein has mitogenic and DNA-binding activity and may play a role in cellular proliferation and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDGF
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HDGF
Gene Accession No. P51858
Gene Alias AI118077; D3Ertd299e; DKFZp686J1764; FLJ96580; Hdgf; Hepatoma-derived growth factor; high mobility group protein 1-like 2; HMG1L2; HMG-1L2; Tdrm1
Gene Symbols HDGF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HDGF. Recombinant protein control fragment (Product #RP-103034).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3068
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.