Learn More
                                    Invitrogen™ HDGF Polyclonal Antibody
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592411
Description
Immunogen sequence: MSRSNRQKEY KCGDLVFAKM KGYPHWPARI DEMPEAAVKS TANKYQVFFF GTHETAFLGP KDLFPYEESK EKFGKPNKRK GFSEGLWEIE NNPTVKASGY QSSQKKSCVE EPEPEPEAAE GDGDKKGNAE GSSDEEGKLV IDEPAKEKNE KGALKRRAGD LLEDSPKRPK EAENPEGEEK EAATLEVERP LPMEVEKNST PSEPGSGRGP PQEEEEEEDE EEEATKEDAE APGIRDHESL; Positive Samples: 22Rv1, HeLa, A-549, 293T, U-251MG, MCF7, BT-474; Cellular Location: Cytoplasm, Nucleus.
HDGF is a member of the hepatoma-derived growth factor family. This protein has mitogenic and DNA-binding activity and may play a role in cellular proliferation and differentiation.
            Specifications
| HDGF | |
| Polyclonal | |
| Unconjugated | |
| HDGF | |
| AI118077; D3Ertd299e; DKFZp686J1764; FLJ96580; Hdgf; Hepatoma-derived growth factor; high mobility group protein 1-like 2; HMG1L2; HMG-1L2; Tdrm1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114499, 15191, 3068 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | 
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P51858, P51859, Q8VHK7 | |
| HDGF | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human HDGF (NP_0044851). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG | 
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.