Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDGFL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564130
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564130 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564130 Supplier Invitrogen™ Supplier No. PA564130
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TAVDEESPFL VAVENGSAPS EPGLVCEPPQ PEE Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 42%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen HDGFL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HDGFL1
Gene Accession No. Q5TGJ6
Gene Alias dJ309H15.1; HDGF (hepatoma-derived growth factor) like; HDGFL1; hepatoma derived growth factor-like 1; hepatoma-derived growth factor-like protein 1; PWWP domain containing 1; PWWP domain-containing protein 1; PWWP1; testicular tissue protein Li 84
Gene Symbols HDGFL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HDGFL1. Recombinant protein control fragment (Product #RP-103523).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154150
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.