Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDGFRP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596696
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596696 Supplier Invitrogen™ Supplier No. PA596696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EELGTLQVTS QILQKNTDVV ATLKKIRRYK ANKDVMEKAA EVYTRLKSRV LGPKIEAVQK VNKAGMEKEK AEEKLAGEEL AGEEAPQEKA EDKPSTDLSA PVNGEATSQK GESAEDKEHE EGRDSEEGPR CGSSEDLHDV REGPDLDRPG SDRQERERAR GDSEALDEES; Positive Samples: Mouse testis; Cellular Location: Nucleus.

Hepatoma-derived growth factor-related protein 2 (HDGFRP2) is a widely expressed member of protein family that contains the HDGF related proteins 1-4 and lens epithelial derived growth factor (LEDGF). These family members show extensive homology at the amino terminus (termed the hath region). Little is known of the role of HDGFRP2, but like the related LEDGF, HDGFRP2 can bind HIV-1 integrase and stimulate HIV-1 integrase activity in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDGFRP2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HDGFL2
Gene Accession No. Q3UMU9, Q7Z4V5
Gene Alias HDGF2; HDGF-2; Hdgf3; HDGF-3; Hdgfl2; HDGF-related protein 2; HDGFRP2; Hepatoma-derived growth factor 2; Hepatoma-derived growth factor 3; hepatoma-derived growth factor, related protein 2; hepatoma-derived growth factor-related protein 2; HRP2; HRP-2; UNQ785/PRO1604
Gene Symbols HDGFL2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 501-670 of human HDGFRP2 (NP_1160201).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15193, 84717
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.