Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HDGFRP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544627
Description
Peptide sequence: GSKRKKSYTS KKSSKQSRKS PGDEDDKDCK EEENKSSSEG GDAGNDTRNT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
HDGFRP3 enhances DNA synthesis and may play a role in cell proliferation.
Specifications
| HDGFRP3 | |
| Polyclonal | |
| Unconjugated | |
| HDGFL3 | |
| 2700022B06Rik; CGI-142; HDGF like 3; HDGF2; HDGF-2; HDGFL3; HDGFRP3; Hepatoma-derived growth factor 2; hepatoma-derived growth factor, related protein 3; hepatoma-derived growth factor-related protein 3; Hrp3; HRP-3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 50810 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y3E1 | |
| HDGFL3 | |
| Synthetic peptide directed towards the middle region of human HDGFRP3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction