Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDGFRP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117150
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117150 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117150 Supplier Invitrogen™ Supplier No. PA5117150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse brain, Mouse kidney, Mouse lung, Rat brain Immunogen sequence: NNPGVKFTGY QAIQQQSSSE TEGEGGNTAD ASSEEEGDRV EEDGKGKRKN EKAGSKRKKS YTSKKSSKQS RKSPGDEDDK DCKEEENKSS SEGGDAGNDT RNTTSDLQKT SEGT.

HDGFRP3 enhances DNA synthesis and may play a role in cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDGFRP3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HDGFL3
Gene Accession No. Q923W4, Q9JMG7, Q9Y3E1
Gene Alias 2700022B06Rik; CGI-142; HDGF like 3; HDGF2; HDGF-2; HDGFL3; HDGFRP3; Hepatoma-derived growth factor 2; hepatoma-derived growth factor, related protein 3; hepatoma-derived growth factor-related protein 3; Hrp3; HRP-3
Gene Symbols HDGFL3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 90-203 of human HDGFRP3 (NP_0571571).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 252941, 29877, 50810
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.