Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDHD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584906
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584906 Supplier Invitrogen™ Supplier No. PA584906
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVEESQTKLK EVFPTAALMP GAEKLIIHLR KHGIPFALAT SSGSASFDMK TSRHKEFFSL FSHIVLGDDP EVQHGEPDPD IFLACAKRFS PPPAMEKCLV FEDAPNGVEA ALAAGMQVVM VPD Highest antigen sequence indentity to the following orthologs: Rat - 73%, Mouse - 68%.

This gene encodes a member of the haloacid dehalogenase-like (HAD) hydrolase superfamily. The encoded protein has no known biological function. This gene has a pseudogene on chromosome 1. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDHD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PUDP
Gene Accession No. Q08623
Gene Alias 1700121L12Rik; 5'-PsiMPase; DXF68S1E; FAM16AX; family with sequence similarity 16, member A, X-linked; GS1; haloacid dehalogenase-like hydrolase domain containing 1; haloacid dehalogenase-like hydrolase domain containing 1A; haloacid dehalogenase-like hydrolase domain-containing protein 1; Haloacid dehalogenase-like hydrolase domain-containing protein 1A; haloacid dehalogenase-like hydrolase domain-containing protein 1A-like; Hdhd1; Hdhd1a; Protein GS1; pseudouridine 5'-phosphatase; pseudouridine-5'-monophosphatase; Pseudouridine-5'-phosphatase; Pudp
Gene Symbols PUDP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PUDP. Recombinant protein control fragment (Product #RP-108603).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8226
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.