Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDHD1 Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA530269
Modifier l'affichage
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIPA530269 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA530269 Fournisseur Invitrogen™ Code fournisseur PA530269
Ajouter au panier
Ajouter au panier

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the haloacid dehalogenase-like (HAD) hydrolase superfamily. The encoded protein has no known biological function. This gene has a pseudogene on chromosome 1. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Spécifications

Antigen HDHD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene PUDP
Gene Accession No. Q08623
Gene Alias 1700121L12Rik; 5'-PsiMPase; DXF68S1E; FAM16AX; family with sequence similarity 16, member A, X-linked; GS1; haloacid dehalogenase-like hydrolase domain containing 1; haloacid dehalogenase-like hydrolase domain containing 1A; haloacid dehalogenase-like hydrolase domain-containing protein 1; Haloacid dehalogenase-like hydrolase domain-containing protein 1A; haloacid dehalogenase-like hydrolase domain-containing protein 1A-like; Hdhd1; Hdhd1a; Protein GS1; pseudouridine 5'-phosphatase; pseudouridine-5'-monophosphatase; Pseudouridine-5'-phosphatase; Pudp
Gene Symbols PUDP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GS1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8226
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats
WARNING: Cancer - www.P65Warnings.ca.gov
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.