Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDHD1A Antibody (OTI7A2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271368F
Description
HDHD1A Monoclonal specifically detects HDHD1A in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
HDHD1A | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
PUDP | |
Full length recombinant protein of human HDHD1A (NP_036212) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG1 |
Flow Cytometry, Immunohistochemistry | |
OTI7A2 | |
Flow Cytometry, Immunohistochemistry | |
DXF68S1E5′-PsiMPase, EC 3.1.3.n6, family with sequence similarity 16, member A, X-linked, GS1FAM16AX, haloacid dehalogenase-like hydrolase domain containing 1, haloacid dehalogenase-like hydrolase domain containing 1A, Haloacid dehalogenase-like hydrolase domain-containing protein 1, Haloacid dehalogenase-like hydrolase domain-containing protein 1A, HDHD1Apseudouridine-5′-monophosphatase, Protein GS1 | |
Mouse | |
Affinity Purified | |
Proteases & Other Enzymes | |
8226.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction