Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDHD2 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299209MFV450
Description
HDHD2 Polyclonal antibody specifically detects HDHD2 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
HDHD2 | |
Polyclonal | |
Western Blot, Immunohistochemistry-Paraffin | |
DKFZp564D1378, DKFZP564D1378,3110052N05Rik, haloacid dehalogenase-like hydrolase domain containing 2, haloacid dehalogenase-like hydrolase domain-containing protein 2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HDHD2. | |
0.1 mL | |
Lipid and Metabolism | |
84064 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction