Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HDHD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559254
Description
Immunogen sequence: TVVGKPEKTF FLEALRGTGC EPEEAVMIGD DCRDDVGGAQ DVGMLGILVK TGKYRASDEE KINPPPYLTC ESFPHAVDH Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
HDHD2 is a protein coding gene.
Specifications
HDHD2 | |
Polyclonal | |
Unconjugated | |
HDHD2 | |
0610039H12Rik; 3110052N05Rik; epididymis secretory protein Li 301; haloacid dehalogenase like hydrolase domain containing 2; haloacid dehalogenase-like hydrolase domain containing 2; haloacid dehalogenase-like hydrolase domain-containing protein 2; hdhd2; HEL-S-301; hypothetical protein LOC505403; hypothetical protein LOC541346; Ier3ip1; ier3ip1 {ECO:0000250; immediate early response 3-interacting protein 1; LRRG00122; RGD1308579; UniProtKB:Q9Y5U9}; zgc:110332 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84064 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H0R4 | |
HDHD2 | |
Recombinant protein corresponding to Human HDHD2. Recombinant protein control fragment (Product #RP-98263). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction