Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDHD2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988459
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8459 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8459 Supplier Bioss Supplier No. BS8317R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HDHD2 is a protein coding gene.

Specifications

Antigen HDHD2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HDHD2
Gene Accession No. Q9H0R4
Gene Alias 0610039H12Rik; 3110052N05Rik; BOS_22245; epididymis secretory protein Li 301; haloacid dehalogenase like hydrolase domain containing 2; haloacid dehalogenase-like hydrolase domain containing 2; Haloacid dehalogenase-like hydrolase domain-containing protein 2; Hdhd2; HEL-S-301; hypothetical protein LOC541346; Ier3ip1; ier3ip1 {ECO:0000250; immediate early response 3-interacting protein 1; LOC100358358; LRRG00122; RGD1308579; UniProtKB:Q9Y5U9}; zgc:110332
Gene Symbols HDHD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HDHD2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84064
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.