Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDHD2B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988461
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8461 Supplier Bioss Supplier No. BS8319R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LHPP is a protein coding gene.

Specifications

Antigen HDHD2B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Lhpp
Gene Accession No. Q9H008
Gene Alias 2310007H09Rik; BOS_23639; HDHD2B; hLHPP; Lhpp; LOC100342924; Phospholysine phosphohistidine inorganic pyrophosphate phosphatase; phospholysine phosphohistidine inorganic pyrophosphate phosphatase-like
Gene Symbols Lhpp
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HDHD2B/LHPP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64077
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.