Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDHD3 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299145V
Description
HDHD3 Polyclonal specifically detects HDHD3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HDHD3 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C9orf158, chromosome 9 open reading frame 158,2810435D12Rik, haloacid dehalogenase-like hydrolase domain containing 3, haloacid dehalogenase-like hydrolase domain-containing protein 3, MGC12904 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human HDHD3. | |
0.1 mL | |
Lipid and Metabolism | |
81932 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction