Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDHD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598981
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598981 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598981 Supplier Invitrogen™ Supplier No. PA598981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HDHD3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

HDHD3 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDHD3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene HDHD3
Gene Accession No. Q9BSH5, Q9CYW4
Gene Alias 2810435D12Rik; C9orf158; haloacid dehalogenase like hydrolase domain containing 3; haloacid dehalogenase-like hydrolase domain containing 3; haloacid dehalogenase-like hydrolase domain-containing protein 3; Hdhd3
Gene Symbols HDHD3
Host Species Rabbit
Immunogen Recombinant Human Haloacid dehalogenase-like hydrolase domain-containing protein 3 protein (1-251AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72748, 81932
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.