Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDJ2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115134
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115134 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115134 Supplier Invitrogen™ Supplier No. PA5115134
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DNAJA1.

The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1. 6 kb and a protein product with a molecular weight of about 35 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDJ2
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene DNAJA1
Gene Accession No. P31689, P63036, P63037
Gene Alias DJ-2; DjA1; DnaJ (Hsp40) homolog, subfamily A, member 1; DnaJ heat shock protein family (Hsp40) member A1; dnaJ homolog subfamily A member 1; DnaJ protein homolog 2; Dnaj2; DNAJA1; dnaJ-like protein 1; DnaJ-like protein 2; DnaJ-like subfamily A member 1; h HSJ-2; HDJ2; hDj-2; Heat shock 40 kDa protein 4; heat shock protein J2; heat shock protein, DNAJ-like 2; HSDJ; HSJ2; HSJ-2; HSPF4; human DnaJ protein 2; I79_019991; LOC100346508; LOW QUALITY PROTEIN: dnaJ homolog subfamily A member 1; NEDD7; neural precursor cell expressed, developmentally down-regulated 7; neural precursor cell expressed, developmentally down-regulated gene 7; Rdj1
Gene Symbols DNAJA1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DNAJA1(Accession P31689), corresponding to amino acid residues P347-S397.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15502, 3301, 65028
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.