Learn More
Invitrogen™ HDJ2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588165
Description
Immunogen sequence: MECQGHGERI SPKDRCKSCN GRKIVREKKI LEVHIDKGMK DGQKITFHGE GDQEPGLEPG DIIIVLDQKD HAVFTRRGED LFMCMDIQLV EALCGFQKPI STLDNRTIVI TSHPGQIVKH GDIKCVLNEG MPIYRRPYEK GRLIIEFKVN FPENGFLSPD KLSLLEKLLP ERKEVEETDE MDQVELVDFD PNQERRRHYN GEAYEDDEHH PRGGVQCQTS; Positive Samples: MCF7, HeLa, Jurkat, U-87MG, HepG2, Mouse brain, Mouse kidney, Mouse liver, Rat liver; Cellular Location: Cytoplasm, Lipid-anchor, Membrane, Microsome, Mitochondrion, Nucleus, perinuclear region.
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1. 6 kb and a protein product with a molecular weight of about 35 kDa.
Specifications
HDJ2 | |
Polyclonal | |
Unconjugated | |
DNAJA1 | |
DJ-2; DjA1; DnaJ (Hsp40) homolog, subfamily A, member 1; DnaJ heat shock protein family (Hsp40) member A1; dnaJ homolog subfamily A member 1; DnaJ protein homolog 2; Dnaj2; DNAJA1; dnaJ-like protein 1; DnaJ-like protein 2; DnaJ-like subfamily A member 1; h HSJ-2; HDJ2; hDj-2; Heat shock 40 kDa protein 4; heat shock protein J2; heat shock protein, DNAJ-like 2; HSDJ; HSJ2; HSJ-2; HSPF4; human DnaJ protein 2; I79_019991; LOC100346508; LOW QUALITY PROTEIN: dnaJ homolog subfamily A member 1; NEDD7; neural precursor cell expressed, developmentally down-regulated 7; neural precursor cell expressed, developmentally down-regulated gene 7; Rdj1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
15502, 3301 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.27 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P31689, P63037 | |
DNAJA1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 178-397 of human DNAJA1 (NP_0015301). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.