Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDLBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582144
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582144 Supplier Invitrogen™ Supplier No. PA582144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DMNQFGEGEQ AKICLEIMQR TGAHLELSLA KDQGLSIMVS GKLDAVMKAR KDIVARLQTQ ASATVAIPKE HHRFVIGKNG EKLQDLELKT ATKIQIPRPD DPSNQIKITG TKEGIEKARH EVLLISAEQD KRAVE Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.

High density lipoprotein-binding protein, also known as vigilin, is a 110-kD protein that specifically binds HDL molecules and may function in the removal of excess cellular cholesterol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDLBP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HDLBP
Gene Accession No. Q00341
Gene Alias 1110005P14Rik; AA960365; AI118566; D1Ertd101e; HBP; HDL binding protein; HDL-binding protein; HDLBP; Hemopexin; high density lipoprotein (HDL) binding protein; high density lipoprotein binding protein; high density lipoprotein binding protein (vigilin); high density lipoprotein-binding protein; Hpx; Hpxn; hx; lipoprotein-binding protein; PRO2900; VGL; vigilin
Gene Symbols HDLBP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HDLBP. Recombinant protein control fragment (Product #RP-102403).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3069
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.